2-(1,3,2-Dioxaborinan-2-yl)phenol 98%
Supplier: Ambeed
|
Warning
|
A1402395-1G
A1402395-5G
76660-548EA
37.14
USD
76660-548
76660-550
2-(1,3,2-Dioxaborinan-2-yl)phenol 98%
2-(1,3,2-Dioxaborinan-2-yl)phenol
Precaución: for Research Use Only
Formula:
C₉H₁₁BO₃ MW: 177.9928 g/mol |
CAS Number:
1121972-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...