13-Hydroxytridecanoic acid 98%
Supplier: Ambeed
|
Warning
|
A1401929-100MG
A1401929-250MG
A1401929-1G
A1401929-5G
76660-544EA
63.66
USD
76660-544
76660-546
77597-142
77622-040
13-Hydroxytridecanoic acid 98%
13-Hydroxytridecanoic acid
Precaución: for Research Use Only
Formula:
C₁₃H₂₆O₃ MW: 230.3437 g/mol |
MDL Number:
MFCD00482032 CAS Number: 7735-38-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...