1-Allylpyrimidine-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A1401705-100MG A1401705-250MG
76660-540EA 55 USD
76660-540 76660-542
1-Allylpyrimidine-2,4(1H,3H)-dione 98%
1-Allylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
CAS Number: 25855-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR