1-Butylpyrimidine-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A1401077-100MG A1401077-250MG
76660-532EA 50 USD
76660-532 76660-534
1-Butylpyrimidine-2,4(1H,3H)-dione 98%
1-Butylpyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MDL Number: MFCD01346711
CAS Number: 705-06-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR