cis-[2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl](2-methylphenyl)nickel(II) chloride 98%

Supplier: Ambeed
Danger

A1399508-100MG A1399508-250MG
76660-530EA 138.98 USD
76660-530 77622-032
cis-[2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl](2-methylphenyl)nickel(II) chloride 98%
cis-[2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl](2-methylphenyl)nickel(II) chloride

Precaución: for Research Use Only

Formula: C₅₁H₃₉ClNiP₂
MW: 807.9493 g/mol
CAS Number: 1594113-05-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR