Cis-[1,2-bis(diphenylphosphino)ethane](2-methylphenyl)nickel(ii) chloride 98%

Supplier: Ambeed
Danger

A1399505-100MG A1399505-1G
76660-528EA 81 USD
76660-528 77549-702
Cis-[1,2-bis(diphenylphosphino)ethane](2-methylphenyl)nickel(ii) chloride 98%
Cis-[1,2-bis(diphenylphosphino)ethane](2-methylphenyl)nickel(ii) chloride

Precaución: for Research Use Only

Formula: C₃₃H₃₁ClNiP₂
MDL Number: MFCD31619187
CAS Number: 1256262-67-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR