tert-Butyl 7-iodoheptanoate 97%

Supplier: Ambeed
Danger

A1393168-1G A1393168-250MG A1393168-5G A1393168-100MG
76660-516EA 79 USD
76660-516 77597-124 77597-126 77622-022
tert-Butyl 7-iodoheptanoate 97%
tert-Butyl 7-iodoheptanoate

Precaución: for Research Use Only

Formula: C₁₁H₂₁IO₂
MW: 312.1877 g/mol
MDL Number: MFCD32642825
CAS Number: 54005-76-4
UN: 2810
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR