5-Isopropyl-1-methyl-1H-pyrazol-3-amine 98%

Supplier: Ambeed
Warning

A1390397-1G A1390397-100MG A1390397-250MG A1390397-5G
76660-508EA 124 USD
76660-508 77867-777 77867-778 77867-779
5-Isopropyl-1-methyl-1H-pyrazol-3-amine 98%
5-Isopropyl-1-methyl-1H-pyrazol-3-amine

Precaución: for Research Use Only

Formula: C₇H₁₃N₃
MDL Number: MFCD26938088
CAS Number: 1229456-20-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR