(R)-(6,6'-Dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphine) 98%

Supplier: Ambeed
Warning

A1386672-100MG A1386672-1G A1386672-250MG
76660-474EA 220 USD
76660-474 76660-476 76660-478
(R)-(6,6'-Dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphine) 98%
(R)-(6,6'-Dimethoxy-[1,1'-biphenyl]-2,2'-diyl)bis(bis(3,5-bis(trimethylsilyl)phenyl)phosphine)

Precaución: for Research Use Only

Formula: C₆₂H₉₆O₂P₂Si₈
CAS Number: 150971-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR