3-Allyl-1-methyl-1H-imidazol-3-ium thiocyanate 99%

Supplier: Ambeed
Warning

A1383545-25G A1383545-5G
76660-460EA 50.93 USD
76660-460 76660-462
3-Allyl-1-methyl-1H-imidazol-3-ium thiocyanate 99%
3-Allyl-1-methyl-1H-imidazol-3-ium thiocyanate

Precaución: for Research Use Only

Formula: C₈H₁₁N₃S
MW: 181.2580 g/mol
MDL Number: MFCD34167267
CAS Number: 861908-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR