exo-Bicyclo[6.1.0]non-4-yn-9-ylmethanol 98%

Supplier: Ambeed
Warning

A1383500-100MG A1383500-250MG A1383500-1G
76660-456EA 113.52 USD
76660-456 76660-458 77622-004
exo-Bicyclo[6.1.0]non-4-yn-9-ylmethanol 98%
exo-Bicyclo[6.1.0]non-4-yn-9-ylmethanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.2176 g/mol
MDL Number: MFCD19705414
CAS Number: 1263291-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR