tert-Butyl 4-(6-chloropyridin-2-yl)piperidine-1-carboxylate 98%

Supplier: Ambeed
Warning

A1381815-1G A1381815-25G
76660-448EA 80.63 USD
76660-448 77566-776
tert-Butyl 4-(6-chloropyridin-2-yl)piperidine-1-carboxylate 98%
tert-Butyl 4-(6-chloropyridin-2-yl)piperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₅H₂₁ClN₂O₂
MW: 296.7924 g/mol
MDL Number: MFCD18206123
CAS Number: 1266118-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR