tert-Butyl (S)-2,7-diazaspiro[4.4]nonane-2-carboxylate 98%

Supplier: Ambeed
Warning

A1380875-1G A1380875-250MG A1380875-5G A1380875-100MG
76660-444EA 272 USD
76660-444 76660-446 77566-774 77867-755
tert-Butyl (S)-2,7-diazaspiro[4.4]nonane-2-carboxylate 98%
tert-Butyl (S)-2,7-diazaspiro[4.4]nonane-2-carboxylate

Precaución: For research use only.

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD31742897
CAS Number: 2306252-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR