2-(2-(((Benzyloxy)carbonyl)amino)ethoxy)ethyl 4-methylbenzenesulfonate 98%

Supplier: Ambeed
Warning

A1379827-100MG A1379827-1G A1379827-250MG A1379827-5G
76660-424EA 11.67 USD
76660-424 76660-426 76660-428 76660-430
2-(2-(((Benzyloxy)carbonyl)amino)ethoxy)ethyl 4-methylbenzenesulfonate 98%
2-(2-(((Benzyloxy)carbonyl)amino)ethoxy)ethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₉H₂₃NO₆S
MW: 393.4540 g/mol
MDL Number: MFCD29059964
CAS Number: 144208-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR