Methyl 2-((tert-butoxycarbonyl)amino)-2-(4,4-difluorocyclohexyl)acetate 98%

Supplier: Ambeed
Warning

A1379708-100MG A1379708-1G A1379708-250MG
76660-418EA 45.62 USD
76660-418 76660-420 76660-422
Methyl 2-((tert-butoxycarbonyl)amino)-2-(4,4-difluorocyclohexyl)acetate 98%
Methyl 2-((tert-butoxycarbonyl)amino)-2-(4,4-difluorocyclohexyl)acetate

Precaución: for Research Use Only

Formula: C₁₄H₂₃F₂NO₄
MW: 307.3335 g/mol
MDL Number: MFCD04115609
CAS Number: 2270904-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR