2-(4-(tert-Butylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1379580-1G A1379580-250MG A1379580-5G
76660-412EA 160.2 USD
76660-412 76660-414 76660-416
2-(4-(tert-Butylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-(tert-Butylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₂₅BO₂S
MW: 292.2445 g/mol
MDL Number: MFCD32197525
CAS Number: 909255-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR