(3-Methyl-4-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid 98%

Supplier: Ambeed
Warning

A1379365-100MG A1379365-1G A1379365-250MG
76660-376EA 37 USD
76660-376 76660-378 76660-380
(3-Methyl-4-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid 98%
(3-Methyl-4-(pyrrolidin-1-ylsulfonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₄S
MDL Number: MFCD31726401
CAS Number: 2377608-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR