2-(4-Chloronaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

A1379163-100MG A1379163-1G A1379163-250MG
76660-364EA 5 USD
76660-364 76660-366 76660-368
2-(4-Chloronaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(4-Chloronaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₆H₁₈BClO₂
MDL Number: MFCD31735828
CAS Number: 2128285-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR