(2-((tert-Butoxycarbonyl)(methyl)amino)pyridin-4-yl)boronic acid 98%

Supplier: Ambeed
Warning

A1379161-100MG A1379161-1G A1379161-250MG
76660-358EA 48 USD
76660-358 76660-360 76660-362
(2-((tert-Butoxycarbonyl)(methyl)amino)pyridin-4-yl)boronic acid 98%
(2-((tert-Butoxycarbonyl)(methyl)amino)pyridin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BN₂O₄
MDL Number: MFCD31726384
CAS Number: 2377609-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR