(6-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-2-methylpyridin-3-yl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A1379119-100MG
A1379119-1G
A1379119-250MG
A1379119-5G
76660-350EA
37
USD
76660-350
76660-352
76660-354
76660-356
(6-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-2-methylpyridin-3-yl)boronic acid 98%
(6-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-2-methylpyridin-3-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₅H₂₄BN₃O₄ |
MDL Number:
MFCD31653729 CAS Number: 2377606-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...