(3aR,3'aR,8aS,8'aS)-8,8a,8',8'a-Tetrahydro-3aH,3'aH-2,2'-biindeno[1,2-d]oxazole ≥97%, ee 99%

Supplier: Ambeed
Warning

A1378974-100MG A1378974-1G A1378974-250MG
76660-332EA 140.04 USD
76660-332 76660-334 76660-336
(3aR,3'aR,8aS,8'aS)-8,8a,8',8'a-Tetrahydro-3aH,3'aH-2,2'-biindeno[1,2-d]oxazole ≥97%, ee 99%
(3aR,3'aR,8aS,8'aS)-8,8a,8',8'a-Tetrahydro-3aH,3'aH-2,2'-biindeno[1,2-d]oxazole

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂O₂
MW: 316.3532 g/mol
CAS Number: 1354558-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR