(3aS,3'aS,8aR,8'aR)-8,8a,8',8'a-Tetrahydro-3aH,3'aH-2,2'-biindeno[1,2-d]oxazole ≥97%, ee 99% (contains ~10% solvents)

Supplier: Ambeed
Warning

A1378972-100MG A1378972-1G A1378972-250MG
76660-326EA 127 USD
76660-326 76660-328 76660-330
(3aS,3'aS,8aR,8'aR)-8,8a,8',8'a-Tetrahydro-3aH,3'aH-2,2'-biindeno[1,2-d]oxazole ≥97%, ee 99% (contains ~10% solvents)
(3aS,3'aS,8aR,8'aR)-8,8a,8',8'a-Tetrahydro-3aH,3'aH-2,2'-biindeno[1,2-d]oxazole

Precaución: for Research Use Only

Formula: C₂₀H₁₆N₂O₂
CAS Number: 1227512-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR