(S)-Ethyl 2-(3,5-dichloro-2-hydroxy-4-(2-(4-methylpiperazin-1-yl)ethoxy)benzamido)-3-phenylpropanoate hydrochloride 98%

Supplier: Ambeed
Warning

A1378782-100MG A1378782-250MG A1378782-50MG
76660-320EA 173 USD
76660-320 76660-322 76660-324
(S)-Ethyl 2-(3,5-dichloro-2-hydroxy-4-(2-(4-methylpiperazin-1-yl)ethoxy)benzamido)-3-phenylpropanoate hydrochloride 98%
(S)-Ethyl 2-(3,5-dichloro-2-hydroxy-4-(2-(4-methylpiperazin-1-yl)ethoxy)benzamido)-3-phenylpropanoate hydrochloride

Precaución: for Research Use Only

Formula: C₂₅H₃₂Cl₃N₃O₅
CAS Number: 2757697-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR