(S)-2-Amino-5-((2,2,10,10-tetramethyl-4,8-dioxo-3,9-dioxa-5,7-diazaundecan-6-ylidene)amino)pentanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: H-Arg(Boc)₂-OH

A1378234-100MG A1378234-250MG A1378234-1G
76660-316EA 69 USD
76660-316 76660-318 77621-972
(S)-2-Amino-5-((2,2,10,10-tetramethyl-4,8-dioxo-3,9-dioxa-5,7-diazaundecan-6-ylidene)amino)pentanoic acid 98%
(S)-2-Amino-5-((2,2,10,10-tetramethyl-4,8-dioxo-3,9-dioxa-5,7-diazaundecan-6-ylidene)amino)pentanoic acid

Precaución: for Research Use Only

Formula: C₁₆H₃₀N₄O₆
CAS Number: 145315-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR