(S)-5-((4-((2-Hydroxy-1-phenylethyl)amino)-5-(1,3,4-oxadiazol-2-yl)pyrimidin-2-yl)amino)-3,3-dimethylisobenzofuran-1(3H)-one 97%

Supplier: Ambeed
Warning

A1378139-100MG A1378139-10MG A1378139-250MG A1378139-25MG A1378139-50MG
76660-304EA 541.06 USD
76660-304 76660-306 76660-308 76660-310 76660-312
(S)-5-((4-((2-Hydroxy-1-phenylethyl)amino)-5-(1,3,4-oxadiazol-2-yl)pyrimidin-2-yl)amino)-3,3-dimethylisobenzofuran-1(3H)-one 97%
(S)-5-((4-((2-Hydroxy-1-phenylethyl)amino)-5-(1,3,4-oxadiazol-2-yl)pyrimidin-2-yl)amino)-3,3-dimethylisobenzofuran-1(3H)-one

Precaución: for Research Use Only

Formula: C₂₄H₂₂N₆O₄
MW: 458.47 g/mol
CAS Number: 2320462-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR