α-(Trifluoromethyl)styrene 97% (by HPLC) stabilized

Supplier: Ambeed
Danger

Synonyms: [1-(Trifluoromethyl)vinyl]benzene, (3,3,3-Trifluoroprop-1-en-2-yl)benzene, Alpha-(trifluoromethyl)styrene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A1378099-100MG A1378099-1G A1378099-250MG
76660-296EA 22.28 USD
76660-296 77549-684 77597-084
α-(Trifluoromethyl)styrene 97% (by HPLC) stabilized
α-(Trifluoromethyl)styrene

Precaución: for Research Use Only

Formula: C₉H₇F₃
MW: 172.147 g/mol
MDL Number: MFCD00081197
CAS Number: 384-64-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR