TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 60% 0.5M in water pH 7.0-7.5

Supplier: Ambeed
Warning

Synonyms: 3,3',3''-Phosphinetriyltripropanoic acid hydrochloride, TCEP, Tris(2-carboxyethyl)phosphine, hydrochloride

A1376686-25G A1376686-500G A1376686-5G
76660-256EA 159.65 USD
76660-256 76660-258 76660-260 76660-262
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 60% 0.5M in water pH 7.0-7.5
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Precaución: for Research Use Only

Formula: C₉H₁₆ClO₆P
MW: 286.65 g/mol
MDL Number: MFCD00145469
CAS Number: 51805-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR