(S)-N-(3-(1-((6-(3,4-Dimethoxyphenyl)pyrazin-2-yl)amino)ethyl)phenyl)-5-methylnicotinamide 98%

Supplier: Ambeed
Warning

A1376658-50MG A1376658-25MG A1376658-100MG
76660-254EA 170 USD
76660-254 77621-958 77867-726
(S)-N-(3-(1-((6-(3,4-Dimethoxyphenyl)pyrazin-2-yl)amino)ethyl)phenyl)-5-methylnicotinamide 98%
(S)-N-(3-(1-((6-(3,4-Dimethoxyphenyl)pyrazin-2-yl)amino)ethyl)phenyl)-5-methylnicotinamide

Precaución: for Research Use Only

Formula: C₂₇H₂₇N₅O₃
MDL Number: MFCD28502207
CAS Number: 1619931-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR