(R)-Methyl 2-amino-3-cyclopentylpropanoate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A1376652-100MG
A1376652-1G
76660-242EA
25
USD
76660-242
76660-244
(R)-Methyl 2-amino-3-cyclopentylpropanoate hydrochloride 97%
(R)-Methyl 2-amino-3-cyclopentylpropanoate hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₈ClNO₂ |
MDL Number:
MFCD33395894 CAS Number: 2514709-11-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...