4-Bromo-1-iododibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A1376459-1G A1376459-250MG A1376459-100MG
76660-234EA 190.97 USD
76660-234 77597-054 77621-956
4-Bromo-1-iododibenzo[b,d]furan 97%
4-Bromo-1-iododibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₁₂H₆BrIO
MW: 372.9839 g/mol
MDL Number: MFCD32215232
CAS Number: 2096506-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR