4-(Prop-2-yn-1-yl)benzonitrile 97%

Supplier: Ambeed
Warning

A1375933-100MG A1375933-1G A1375933-250MG
76660-222EA 131 USD
76660-222 76660-224 76660-226
4-(Prop-2-yn-1-yl)benzonitrile 97%
4-(Prop-2-yn-1-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇N
MDL Number: MFCD18250553
CAS Number: 1260663-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR