4-Bromo-2-ethyl-1-pentylbenzene 97%

Supplier: Ambeed
Warning

A1375562-100MG A1375562-250MG
76660-214EA 127.31 USD
76660-214 76660-216
4-Bromo-2-ethyl-1-pentylbenzene 97%
4-Bromo-2-ethyl-1-pentylbenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₉Br
MW: 255.1900 g/mol
MDL Number: MFCD34165700
CAS Number: 2703752-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR