1,5-Bis(4-bromophenyl)pentan-3-one 97%

Supplier: Ambeed
Warning

A1375491-100MG A1375491-1G A1375491-250MG A1375491-5G
76660-206EA 29.71 USD
76660-206 76660-208 76660-210 76660-212
1,5-Bis(4-bromophenyl)pentan-3-one 97%
1,5-Bis(4-bromophenyl)pentan-3-one

Precaución: for Research Use Only

Formula: C₁₇H₁₆Br₂O
MW: 396.1163 g/mol
MDL Number: MFCD09836436
CAS Number: 937635-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR