Benzo[b][1,4]dioxine-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A1375479-100MG A1375479-1G A1375479-250MG
76660-200EA 162 USD
76660-200 76660-202 76660-204
Benzo[b][1,4]dioxine-6-carboxylic acid 97%
Benzo[b][1,4]dioxine-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆O₄
MW: 178.1415 g/mol
CAS Number: 67470-99-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR