Ethyl 1-methyl-4-(trifluoromethyl)-1H-pyrazole-3-carboxylate 97%

Supplier: Ambeed
Warning

A1375439-100MG A1375439-1G
76660-196EA 203.7 USD
76660-196 76660-198
Ethyl 1-methyl-4-(trifluoromethyl)-1H-pyrazole-3-carboxylate 97%
Ethyl 1-methyl-4-(trifluoromethyl)-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₈H₉F₃N₂O₂
MW: 222.1645 g/mol
CAS Number: 197226-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR