(R)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine 98%

Supplier: Ambeed
Warning

Synonyms: (R)-(R)-PPFA

A1375357-1G A1375357-250MG A1375357-100MG
76660-180EA 114.58 USD
76660-180 76660-182 77621-952
(R)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine 98%
(R)-N,N-Dimethyl-1-[(R)-2-(diphenylphosphino)ferrocenyl]ethylamine

Precaución: for Research Use Only

Formula: C₂₆H₂₈FeNP
MW: 441.34 g/mol
MDL Number: MFCD00075286
CAS Number: 55700-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR