((4-Hydroxybutyl)azanediyl)bis(hexane-6,1-diyl) bis(2-hexyldecanoate) 97%

Supplier: Ambeed
Warning

A1375212-100MG A1375212-1G A1375212-250MG A1375212-25MG A1375212-50MG A1375212-10MG
76660-154EA 270 USD
76660-154 76660-156 76660-158 76660-160 76660-162 77621-944
((4-Hydroxybutyl)azanediyl)bis(hexane-6,1-diyl) bis(2-hexyldecanoate) 97%
((4-Hydroxybutyl)azanediyl)bis(hexane-6,1-diyl) bis(2-hexyldecanoate)

Precaución: for Research Use Only

Formula: C₄₈H₉₅NO₅
MDL Number: MFCD33402175
CAS Number: 2036272-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR