Indane 99%

Supplier: Ambeed
Danger

Synonyms: 2,3-Dihydro-1H-indene, Indan

A1375172-25G A1375172-5G
76660-152EA 18 USD
76660-152 77566-750
Indane 99%
Indane

Precaución: For research use only.

Formula: C₉H₁₀
MW: 118.18 g/mol
Boiling Pt: 176.5
Melting Pt: -51.4
MDL Number: MFCD00003795
CAS Number: 496-11-7
UN: 3295
ADR: 3 Ⅲ
Merck Index: 13,04956

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR