N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 97% (by GC)

Supplier: Ambeed
Warning

A1375120-100G A1375120-25G A1375120-500G
76660-148EA 34 USD
76660-148 76660-150 77549-678
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine 97% (by GC)
N,N,N',N'-Tetrakis(2-hydroxyethyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₁₀H₂₄N₂O₄
MDL Number: MFCD00002854
CAS Number: 140-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR