N1,N2-Bis(naphthalen-1-ylmethyl)oxalamide 90%

Supplier: Ambeed
Danger

A1374787-100MG A1374787-1G A1374787-250MG A1374787-5G
76660-144EA 126 USD
76660-144 76660-146 77592-608 77867-706
N1,N2-Bis(naphthalen-1-ylmethyl)oxalamide 90%
N1,N2-Bis(naphthalen-1-ylmethyl)oxalamide

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₂O₂
MDL Number: MFCD32633515
CAS Number: 2281918-10-1
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR