7-Bromo-1H-indole-6-carbonitrile 98%
Supplier: Ambeed
|
Warning
|
A1374663-5G
A1374663-100MG
A1374663-1G
A1374663-250MG
76660-136EA
1469.35
USD
76660-136
77592-606
77597-046
77597-048
7-Bromo-1H-indole-6-carbonitrile 98%
7-Bromo-1H-indole-6-carbonitrile
Precaución: for Research Use Only
Formula:
C₉H₅BrN₂ MW: 221.05 g/mol |
MDL Number:
MFCD34179910 CAS Number: 2173193-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...