L(+)-Rhamnose 97% (contains < 9-12% H₂O)

Supplier: Ambeed
Warning

Synonyms: 6-Deoxy-L-mannose, 6-Deoxy-L(+)-mannose, (2R,3R,4S,5S)-2,3,4,5-Tetrahydroxyhexanal

A1374313-100G A1374313-25G A1374313-500G
76660-130EA 33 USD
76660-130 76660-132 76660-134
L(+)-Rhamnose 97% (contains < 9-12% H₂O)
L(+)-Rhamnose

Precaución: for Research Use Only

Formula: C₆H₁₂O₅
MW: 164.16
Melting Pt: 94
Density: 1.47
MDL Number: MFCD00066373
CAS Number: 3615-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR