2-Bromo-5-chloro-6-methylpyridin-3-amine 97%

Supplier: Ambeed
Warning

A1374307-100MG A1374307-1G A1374307-250MG A1374307-5G
76660-122EA 135 USD
76660-122 76660-124 76660-126 76660-128
2-Bromo-5-chloro-6-methylpyridin-3-amine 97%
2-Bromo-5-chloro-6-methylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₆H₆BrClN₂
MW: 221.4822 g/mol
MDL Number: MFCD28952942
CAS Number: 1936218-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR