(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]-2-[(1R)-1-[bis(3,5-dimethylphenyl)phosphino]ethyl]ferrocene 95%

Supplier: Ambeed
Warning

A1374277-100MG A1374277-1G A1374277-250MG
76660-100EA 34 USD
76660-100 76660-102 76660-104
(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]-2-[(1R)-1-[bis(3,5-dimethylphenyl)phosphino]ethyl]ferrocene 95%
(1S)-1-[Bis(3,5-dimethylphenyl)phosphino]-2-[(1R)-1-[bis(3,5-dimethylphenyl)phosphino]ethyl]ferrocene

Precaución: for Research Use Only

CAS Number: 158923-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR