Methyl 7-bromo-3-hydroxy-2-naphthoate 97%

Supplier: Ambeed
Warning

A1374259-1G A1374259-25G A1374259-5G
76660-094EA 24.41 USD
76660-094 76660-096 76660-098
Methyl 7-bromo-3-hydroxy-2-naphthoate 97%
Methyl 7-bromo-3-hydroxy-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₂H₉BrO₃
MW: 281.1021 g/mol
MDL Number: MFCD25970084
CAS Number: 10155-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR