5-Iodo-2,3,3-trimethyl-3H-indole 97%

Supplier: Ambeed
Warning

A1373886-100MG A1373886-1G A1373886-250MG A1373886-5G
76660-076EA 23 USD
76660-076 76660-078 76660-080 76660-082
5-Iodo-2,3,3-trimethyl-3H-indole 97%
5-Iodo-2,3,3-trimethyl-3H-indole

Precaución: for Research Use Only

Formula: C₁₁H₁₂IN
MW: 285.1242 g/mol
MDL Number: MFCD25964381
CAS Number: 54136-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR