(S)-2,2-Dimethylchroman-4-amine hydrochloride 97%

Supplier: Ambeed
Warning

A1373806-100MG A1373806-250MG
76660-070EA 75 USD
76660-070 76660-072
(S)-2,2-Dimethylchroman-4-amine hydrochloride 97%
(S)-2,2-Dimethylchroman-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₆ClNO
MDL Number: MFCD34165695
CAS Number: 2646585-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR