(S)-2-(Methylamino)butanamide hydrochloride 97%

Supplier: Ambeed
Warning

A1373752-100MG A1373752-1G A1373752-250MG
76660-064EA 68.96 USD
76660-064 76660-066 76660-068
(S)-2-(Methylamino)butanamide hydrochloride 97%
(S)-2-(Methylamino)butanamide hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₃ClN₂O
MW: 152.6225 g/mol
MDL Number: MFCD33395875
CAS Number: 2682097-15-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR