Bis(2-((4R,5R)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)phenyl)amine 98% +99%ee

Supplier: Ambeed
Warning

A1373670-100MG A1373670-1G A1373670-250MG
76660-058EA 220 USD
76660-058 76660-060 76660-062
Bis(2-((4R,5R)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)phenyl)amine 98% +99%ee
Bis(2-((4R,5R)-4,5-diphenyl-4,5-dihydrooxazol-2-yl)phenyl)amine

Precaución: for Research Use Only

Formula: C₄₂H₃₃N₃O₂
CAS Number: 1370549-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR