(9H-Fluoren-9-yl)methyl (3-bromophenyl)carbamate 97%

Supplier: Ambeed
Warning

A1373564-250MG A1373564-5G
76660-052EA 10 USD
76660-052 77867-696
(9H-Fluoren-9-yl)methyl (3-bromophenyl)carbamate 97%
(9H-Fluoren-9-yl)methyl (3-bromophenyl)carbamate

Precaución: for Research Use Only

Formula: C₂₁H₁₆BrNO₂
MW: 394.2612 g/mol
MDL Number: MFCD30626242
CAS Number: 2152776-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR